Order Entry
United States
ContactUsLinkComponent
 
Anti-CACNG3 Rabbit Polyclonal Antibody
  103338-744
Anti-CACNG3 Rabbit Polyclonal Antibody
  103338-744
 :  H00010368-D01P

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CACNG3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    10368
  • Antigen Synonyms:
    gamma subunit 3|Neuronal voltage-gated calcium channel gamma-3 subunit|calcium channel|voltage-dependent calcium channel gamma-3 subunit|voltage-gated calcium channel gamma subunit|voltage-dependent
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    CACNG3 (NP_006530.1, 1 a.a. ~ 315 a.a) full-length human protein.
  • Purification:
    Protein G purified
  • Cat. No.:
    103338-744
  • Supplier no.:
    H00010368-D01P

 

 

The CACNG3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CACNG3. This antibody reacts with human. The CACNG3 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CACNG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human