Anti-CACNA1S Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1A]
103394-924
- Antibody Type:Primary
- Antigen Symbol:CACNA1S
- Clonality:Monoclonal
- Clone:1A
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Guinea pig,Rat,Mouse
- Size:0.2 ml
- Gene ID:779
- Antigen Synonyms:isoform 3|hypokalemic periodic paralysis)|skeletal muscle|Cav1.1|calcium channel|CACH1|CACN1|CACNL1A3MHS5|L type|voltage-dependent L-type calcium channel subunit alpha-1S|Calcium channel|TTPP1|HOKPP|HOKPP1|hypoPP|isoform 3 (skeletal muscle|dihydropyridine-sensitive L-type calcium channel alpha-1 subunit|alpha 1 polypeptide|Voltage-gated calcium channel subunit alpha Cav1.1|alpha-1 polypeptide|CCHL1A3|voltage-dependent|dihydropyridine receptor|alpha 1S subunit
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Purified rabbit muscle T-tubule dihydropyridine receptor.
- Purification:Protein A purified
- Cat. No.:103394-924
The CACNA1S Antibody (1A) [FITC] from Novus Biologicals is a mouse monoclonal antibody to CACNA1S. This antibody reacts with human, mouse, rat, guinea pig, rabbit. The CACNA1S Antibody (1A) [FITC] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: CACNA1S
Clonality: Monoclonal
Clone: 1A
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Guinea Pig, Rabbit