Anti-CA11 Mouse Polyclonal Antibody
103322-988
- Antibody Type:Primary
- Antigen Symbol:CA11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:770
- Antigen Synonyms:carbonic anhydrase XI|carbonic anhydrase-related protein XI|CARP-2|carbonic anhydrase-related protein 11|Carbonic anhydrase-related protein 2|CARPX1|CA-RP II|CARP2CARP XI|CA-XI|CA-RP XI
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CA11 (NP_001208.2, 1 a.a. - 328 a.a.) full-length human protein. MGAAARLSAPRALVLWAALGAAAHIGPAPDPEDWWSYKDNLQGNFVPGPPFWGLVNAAWSLCAVGKRQSPVDVELKRVLYDPFLPPLRLSTGGEKLRGTLYNTGRHVSFLPAPRPVVNVSGGPLLYSHRLSELRLLFGARDGAGSEHQINHQGFSAEVQLIHFNQELYGNFSAASRGPNGLAILSLFVNVASTSNPFLSRLLNRDTITRISYKNDAYFLQDLSLELLFPESFGFITYQGSLSTPPCSETVTWILIDRALNITSLQMHSLRLLSQNPPSQIFQSLSGNSRPLQPLAHRALRGNRDPRHPERRCRGPNYRLHVDGVPHGR
- Purification:Protein A purified
- Cat. No.:103322-988
- Supplier no.:H00000770-B02P
The CA11 Antibody from Novus Biologicals is a mouse polyclonal antibody to CA11. This antibody reacts with human. The CA11 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human