Anti-C2orf62 Mouse Monoclonal Antibody [clone: 2C3]
103348-390
- Antibody Type:Primary
- Antigen Symbol:C2orf62
- Clonality:Monoclonal
- Clone:2C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:375307
- Antigen Synonyms:hypothetical protein LOC375307|MGC50811|chromosome 2 open reading frame 62
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MGC50811 (AAH52750, 1 a.a. ~ 388 a.a) full length recombinant protein with GST tag.MSSKVYSTGSRAKDHQPSGPECLPLPEANAEAIDFLSSLHKEELQMLFFSETLAMVSDTGEPQGELTIEVQRGKYQEKLGMLTYCLFVHASSRGFLDKMLCGNSLLGYLSEKLELMEQHSQDFIKFLILPMERKMSLLKQDDQLAVTRSIKEGEEVKTGVTSFPWSSIKGFISEAANLVLLRVMAWRRMVPSNARFLTLDTEGKLCYLTYQNLGFQTIQVDHQQAEVFIVEQTVHAEEGIPMSCQYYLLSDGHLA
- Purification:IgG purified
- Cat. No.:103348-390
- Supplier no.:H00375307-M01
The C2orf62 Antibody (2C3) from Novus Biologicals is a mouse monoclonal antibody to C2orf62. This antibody reacts with human. The C2orf62 Antibody (2C3) has been validated for the following applications: ELISA.
Type: Primary
Antigen: C2orf62
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human