Anti-BXDC5 Rabbit Polyclonal Antibody
103279-968
- Antibody Type:Primary
- Antigen Symbol:BXDC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:80135
- Antigen Synonyms:RNA processing factor 1 (RPF1)|brix domain containing 5|BXDC5|ribosome production factor 1|DKFZp761M0215|Brix domain-containing protein 5|BBR140|RP11-118B23.1|ribosome production factor 1 homolog (S. cerevisiae)|Ribosome biogenesis protein RPF1|FLJ12475|2310066N05Rik|DKFZp761G0415
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:SGKKSLKRKAAAEELQEAAGAGDGATENGVQPPKAAAFPPGFSISEIKNKQRRHLMFTRWKQQQRKEKLAAKKKLKKEREALGDKAPPKPVPKTID
- Purification:Immunogen affinity purified
- Cat. No.:103279-968
- Supplier no.:NBP1-89915
The BXDC5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BXDC5. This antibody reacts with human, mouse, rat. The BXDC5 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: BXDC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat