Anti-BMP-8b Mouse Monoclonal Antibody (DyLight 650) [clone: 13E6]
103393-860
- Antibody Type:Primary
- Antigen Symbol:BMP-8b
- Clonality:Monoclonal
- Clone:13E6
- Conjugation:DyLight® 650
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:656
- Antigen Synonyms:BMP-8|MGC131757|bone morphogenetic protein 8b
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human BMP8b (264-402aa) purified from E. coli
- Purification:Protein A purified
- Cat. No.:103393-860
The BMP-8b Antibody (13E6) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to BMP-8b. This antibody reacts with human. The BMP-8b Antibody (13E6) [DyLight 650] has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: BMP-8b
Clonality: Monoclonal
Clone: 13000000
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human