Anti-BMP-2 Mouse Monoclonal Antibody (DyLight 680) [clone: 15B3]
Catalog # 103393-836
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:BMP-2
- Clonality:Monoclonal
- Clone:15B3
- Conjugation:DyLight® 680
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:650
- Antigen Synonyms:BMP2ABone morphogenetic protein 2A|BMP-2|bone morphogenetic protein 2|BMP-2A
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human BMP2 (283-396aa) purified from E. coli
- Purification:Protein A purified
- Cat. No.:103393-836
Specifications
About this item
The BMP-2 Antibody (15B3) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to BMP-2. This antibody reacts with human. The BMP-2 Antibody (15B3) [DyLight 680] has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: BMP-2
Clonality: Monoclonal
Clone: 15B3
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human