Anti-BIN1 Mouse Monoclonal Antibody [clone: 1H1]
Catalog # 103321-786
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:BIN1
- Clonality:Monoclonal
- Clone:1H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:274
- Antigen Synonyms:Amphiphysin-like protein|box dependant MYC interacting protein 1|myc box-dependent-interacting protein 1|Amphiphysin II|AMPH2|SH3P9|MGC10367|bridging integrator 1Box-dependent myc-interacting protein 1|AMPHLDKFZp547F068
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:BIN1 (NP_004296 355 a.a. - 454 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VVETFPATVNGTVEGGSGAGRLDLPPGFMFKVQAQHDYTATDTDELQLKAGDVVLVIPFQNPEEQDEGWLMGVKESDWNQHKELEKCRGVFPENFTERVP
- Purification:IgG purified
- Cat. No.:103321-786
- Supplier no.:H00000274-M01
Specifications
About this item
The BIN1 Antibody (1H1) from Novus Biologicals is a mouse monoclonal antibody to BIN1. This antibody reacts with human. The BIN1 Antibody (1H1) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 1H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human