Anti-BDH1 Mouse Polyclonal Antibody
Catalog # 103322-558
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:BDH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:622
- Antigen Synonyms:mitochondrial)|3-hydroxybutyrate dehydrogenase|type 1|(R)-3-hydroxybutyrate dehydrogenase|short chain dehydrogenase/reductase family 9C|mitochondrial|member 1|EC 1.1.1|MGC9788|MGC4347|MGC2723
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:BDH1 (NP_004042.1, 1 a.a. - 343 a.a.) full-length human protein. MLATRLSRPLSRLPGKTLSACDRENGARRPLLLGSTSFIPIGRRTYASAAEPVGSKAVLVTGCDSGFGFSLAKHLHSKGFLVFAGCLMKDKGHDGVKELDSLNSDRLRTVQLNVCSSEEVEKVVEIVRSSLKDPEKGMWGLVNNAGISTFGEVEFTSLETYKQVAEVNLWGTVRMTKSFLPLIRRAKGRVVNISSMLGRMANPARSPYCITKFGVEAFSDCLRYEMYPLGVKVSVVEPGNFIAATSLYSPESIQAIAKKMWEELPEVVRKDYGKKYFDEKIAKMETYCSSGSTDTSPVIDAVTHALTATTPYTRYHPMDYYWWLRMQIMTHLPGAISDMIYIR
- Purification:Protein A purified
- Cat. No.:103322-558
- Supplier no.:H00000622-B01P
Specifications
About this item
The BDH1 Antibody from Novus Biologicals is a mouse polyclonal antibody to BDH1. This antibody reacts with human. The BDH1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: BDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human