Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Breast cancer suppressor candidate 1
- Antigen Symbol:BCSC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4013
- Antigen Synonyms:LOH11CR2Avon Willebrand factor A domain-containing protein 5A|BCSC1ortholog of mouse AW551984|11|loss of heterozygosity|gene A|von Willebrand factor A domain containing 5A|chromosomal region 2|BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein|Breast cancer suppressor candidate 1
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human LOH11CR2A. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102706-332
- Supplier no.:NBP2-15615
Specifications
About this item
The Breast cancer suppressor candidate 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Breast cancer suppressor candidate 1. This antibody reacts with human. The Breast cancer suppressor candidate 1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: BCSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human