To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:BCAP/PIK3AP1
- Clonality:Monoclonal
- Clone:3C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:118788
- Antigen Synonyms:1810044J04Rik|phosphoinositide 3-kinase adapter protein 1|RP11-34E5.3|phosphoinositide-3-kinase adaptor protein 1|B cell adaptor protein|FLJ35564|BCAPB-cell adapter for phosphoinositide 3-kinase|B-cell phosphoinositide 3-kinase adapter protein 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PIK3AP1 (NP_689522, 601 a.a. ~ 701 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103346-468
- Supplier no.:H00118788-M01
Specifications
About this item
The BCAP / PIK3AP1 Antibody (3C2) from Novus Biologicals is a mouse monoclonal antibody to BCAP / PIK3AP1. This antibody reacts with human. The BCAP / PIK3AP1 Antibody (3C2) has been validated for the following applications: ELISA.
Type: Primary
Antigen: BCAP/PIK3AP1
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human