Anti-Band 3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Q1/156]
Catalog # 103403-376
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Band 3
- Antigen Symbol:Band 3
- Clonality:Monoclonal
- Clone:Q1/156
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Gene ID:6521
- Antigen Synonyms:BND3|erythrocyte membrane protein band 3|RTA1A|AE 1|FR|Solute carrier family 4 member 1|Wright blood group|Waldner blood group|WD1|Froese blood group|Swann blood group|CD233|Anion exchanger 1|EMPB3|MGC116750|EPB3MGC126623|number 1|SW|CD233 antigen|member 1 (erythrocyte membraneprotein band 3|MGC116753|erythroid anion exchange protein|DI|WD|Anion exchange protein 1|Diego blood group)|solute carrier family 4|band 3 anion transport protein|anion exchanger|AE1MGC126619|WR
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Foetal liver cells
- Purification:Protein G purified
- Cat. No.:103403-376
Specifications
About this item
The Band 3 Antibody (Q1 / 156) [FITC] from Novus Biologicals is a mouse monoclonal antibody to Band 3. This antibody reacts with human. The Band 3 Antibody (Q1 / 156) [FITC] has been validated for the following applications: Immunohistochemistry.
Type: Primary
Antigen: Band 3
Clonality: Monoclonal
Clone: Q1/156
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human