Anti-BAAT Mouse Monoclonal Antibody [clone: 1E4.]
Catalog # 103322-484
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:BAAT
- Clonality:Monoclonal
- Clone:1E4.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:570
- Antigen Synonyms:MGC104432|BACAT|EC 2.3.1.65|bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase)|FLJ20300|bile acid Coenzyme A: amino acid N-acyltransferase (glycineN-choloyltransferase)|Long-chain fatty-acyl-CoA hydrolase|Glycine N-choloyltransferase|BATbile acid-CoA:amino acid N-acyltransferase|EC 3.1.2.2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:BAAT (NP_001692 258 a.a. - 355 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NGTNFPFGIPQVYHGQIHQPLPHSAQLISTNALGLLELYRTFETTQVGASQYLFPIEEAQGQFLFIVGEGDKTINSKAHAEQAIGQLKRHGKNNWTLL
- Purification:IgG purified
- Cat. No.:103322-484
- Supplier no.:H00000570-M03
Specifications
About this item
The BAAT Antibody (1E4.) from Novus Biologicals is a mouse monoclonal antibody to BAAT. This antibody reacts with human. The BAAT Antibody (1E4.) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: BAAT
Clonality: Monoclonal
Clone: 1E4.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human