Anti-B3GNT3 Mouse Polyclonal Antibody
Catalog # 103338-666
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:B3GNT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:10331
- Antigen Synonyms:Beta-1,3-Gn-T3|Beta3Gal-T8|TMEM3beta3Gal-T8|UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8|Beta-1,3-galactosyltransferase 8|HP10328|BGnT-3|B3GN-T3|core1-beta3GlcNAcT|B3GAL-T8|UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8|B3GNT-3|Beta-3-Gx-T8|transmembrane protein 3|putative type II membrane protein|B3GALT8|Beta3GalT8|EC 2.4.1.-|EC 2.4.1|beta-1,3-N-acetylglucosaminyltransferase bGnT-3|UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3|beta3Gn-T3|Beta-1,3-GalTase 8|Core 1 extending beta-1,3-N-acetylglucosaminyltransferase|Beta-1,3-N-acetylglucosaminyltransferase 3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:B3GNT3 (AAH67423.1, 1 a.a. ~ 372 a.a) full-length human protein.MKYLRHRRPNATLILAIGAFTLLLFSLLVSPPTCKVQEQPPAIPEALAWPTPPTRPAPAPCHANTSMVTHPDFATQPQHVQNFLLYRHCRHFPLLQDVPPSKCAQPVFLLLVIKSSPSNYVRRELLRRTWGRERKVRGLQLRLLFLVGTASNPHEARKVNRLLELEAQTHGDILQWDFHDSFFNLTLKQVLFLQWQETRCANASFVLNGDDDVFAHTDNMVFYLQDHDPGRHLFVGQLIQNVGPIRAFWSKYYVP
- Purification:Protein A purified
- Cat. No.:103338-666
- Supplier no.:H00010331-B01P
Specifications
About this item
The B3GNT3 Antibody from Novus Biologicals is a mouse polyclonal antibody to B3GNT3. This antibody reacts with human. The B3GNT3 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: B3GNT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human