Anti-AUF1 Rabbit Polyclonal Antibody
Catalog # 103328-300
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:AUF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:3184
- Antigen Synonyms:hnRNPD0|heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein1|AUF1A|heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA-binding protein1|heterogeneous nuclear ribonucleoprotein D0|37kDa)|37kD)|type A
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:HNRNPD (NP_112738.1, 1 a.a. - 355 a.a.) full-length human protein. MSEEQFGGDGAAAAATAAVGGSAGEQEGAMVAATQGAAAAAGSGAGTGGGTASGGTEGGSAESEGAKIDASKNEEDEGHSNSSPRHSEAATAQREEWKMFIGGLSWDTTKKDLKDYFSKFGEVVDCTLKLDPITGRSRGFGFVLFKESESVDKVMDQKEHKLNGKVIDPKRAKAMKTKEPVKKIFVGGLSPDTPEEKIREYFGGFGEVESIELPMDNKTNKRRGFCFITFKEEEPVKKIMEKKYHNVGLSKCEIKVAMSKEQYQQQQQWGSRGGFAGRARGRGGGPSQNWNQGYSNYWNQGYGNYGYNSQGYGGYGGYDYTGYNNYYGYGDYSNQQSGYGKVSRRGGHQNSYKPY
- Purification:Protein A purified
- Cat. No.:103328-300
- Supplier no.:H00003184-D01P
Specifications
About this item
The AUF1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AUF1. This antibody reacts with human. The AUF1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: AUF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human