To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ATRX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:546
- Antigen Synonyms:alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog)|alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog|RAD54 homolog|EC 3.6.4.12|XNPZNF-HX|alpha thalassemia/mental retardation syndrome X-linked|X-linked|Znf-HX|DNA dependent ATPase and helicase|ATR2|RAD54L|SFM1|Juberg-Marsidi syndrome|EC 3.6.1|Zinc finger helicase|SHS|XH2RAD54|X-linked helicase II|ATP-dependent helicase ATRX|helicase 2|MRXHF1|S.cerevisiae)|MGC2094|transcriptional regulator ATRX|X-linked nuclear protein
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:AAWAEYEAEKKGLTMRFNIPTGTNLPPVSFNSQTPYIPFNLGALSAMSNQQLEDLINQGREKVVEATNSVTAVRIQPLEDIISAVWKENMNLSEAQVQALALSRQASQELDVKRREAIYNDVLTKQQMLISCVQRILMNRR
- Purification:Immunogen affinity purified
- Cat. No.:103267-320
- Supplier no.:NBP1-83077
Specifications
About this item
The ATRX Antibody from Novus Biologicals is a rabbit polyclonal antibody to ATRX. This antibody reacts with human. The ATRX Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ATRX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human