Anti-ATG4A Goat Polyclonal Antibody
Catalog # 103405-882
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ATG4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Gene ID:115201
- Antigen Synonyms:EC 3.4.22|APG4 autophagy 4 homolog A (S. cerevisiae)|APG4A|AUT-like 2 cysteine endopeptidase|EC 3.4.22.-|cysteine endopeptidase (S. cerevisiae)|ATG4 autophagy related 4 homolog A (S. cerevisiae)|AUT-like 2|autophagin-2|hAPG4A|autophagin 2|cysteine protease ATG4A|AUTL2|cysteine endopeptidase
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence TEENGTVNDQTFHC corresponding to internal region according to NP_443168.2, NP_840054.1.
- Purification:Immunogen affinity purified
- Cat. No.:103405-882
Specifications
About this item
The ATG4A Antibody from Novus Biologicals is a goat polyclonal antibody to ATG4A. This antibody reacts with human. The ATG4A Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: ATG4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human