To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ATF2
- Clonality:Monoclonal
- Clone:103C411.2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:1386
- Antigen Synonyms:CREBP1|cAMP-responsive element-binding protein 2|activating transcription factor 2formerly|cyclic AMP-dependent transcription factor ATF-2|MGC111558|cAMP responsive element binding protein 2
- Modification:p Thr71
- Storage Buffer:PBS - BSA free
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was generated by immunizing mice with a synthetic peptide containing phosphorylated threonine at position 71 of human ATF-2.
- Purification:Protein G purified
- Cat. No.:103401-802
Specifications
About this item
The ATF2 [p Thr71] Antibody (103C411.2) - Azide Free from Novus Biologicals is a mouse monoclonal antibody to ATF2. This antibody reacts with human. The ATF2 [p Thr71] Antibody (103C411.2) - Azide Free has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: ATF2
Clonality: Monoclonal
Clone: 103C411.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human