Anti-ARMCX3 Rabbit Polyclonal Antibody
Catalog # 103271-334
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ARMCX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:51566
- Antigen Synonyms:ALEX3arm protein lost in epithelial cancers|armadillo repeat-containing X-linked protein 3,1200004E24Rik|dJ545K15.2|armadillo repeat containing|Protein ALEX3|MGC12199|X-linked 3|KIAA0443|ARM protein lost in epithelial cancers on chromosome X 3|DKFZp781N1954|X chromosome|3
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:SAGNEETKLQVLKLLLNLAENPAMTRELLRAQVPSSLGSLFNKKENKEVILKLLVIFENINDNFKWEENEPTQNQFGEGSLFFFLKEFQVCADKVLGIESHHDFLVKVKVGKFMAKLAEHM
- Purification:Immunogen affinity purified
- Cat. No.:103271-334
- Supplier no.:NBP1-89142
Specifications
About this item
The ARMCX3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ARMCX3. This antibody reacts with human. The ARMCX3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ARMCX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human