Anti-ARID5B Mouse Monoclonal Antibody [clone: 1C2]
Catalog # 103345-080
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ARID5B
- Clonality:Monoclonal
- Clone:1C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:84159
- Antigen Synonyms:modulator recognition factor 2 (MRF2)|DESRTAT-rich interactive domain-containing protein 5B|ARID domain-containing protein 5B|Modulator recognition factor 2|MRF2MRF1-like protein|AT rich interactive domain 5B (MRF1-like)|FLJ21150|MRF-2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ARID5B (XP_084482.3, 1483 a.a. ~ 1583 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.SGLNSRLPAGYSHSLQYLKNQTVLSPLMQPLAFHSLVMQRGIFTSPTNSQQLYRHLAAATPVGSSYGDLLHNSIYPLAAINPQAAFPSSQLSSVHPSTKL
- Purification:IgG purified
- Cat. No.:103345-080
- Supplier no.:H00084159-M04
Specifications
About this item
The ARID5B Antibody (1C2) from Novus Biologicals is a mouse monoclonal antibody to ARID5B. This antibody reacts with human. The ARID5B Antibody (1C2) has been validated for the following applications: ELISA.
Type: Primary
Antigen: ARID5B
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human