To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ARID1B
- Clonality:Monoclonal
- Clone:2D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:57492
- Antigen Synonyms:PLU1|JPUT1|OSA2|p250R|KIAA1235AT-rich interactive domain-containing protein 1B|BRG1-binding protein ELD/OSA1|ELD (eyelid)/OSA protein|BAF250B|AT rich interactive domain 1B (SWI1-like)|RBBP2H1A|hOsa2|6A3-5|ELD/OSA1|Osa homolog 2|KDM5B|DAN15P250R|BRG1-binding protein hELD/OSA1|BRG1-associated factor 250b|BRIGHT|ARID domain-containing protein 1B
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ARID1B (NP_059989, 1364 a.a. ~ 1461 a.a) partial recombinant protein with GST tag.PPAKRHEGDMYNMQYSSQQQEMYNQYGGSYSGPDRRPIQGQYPYPYSRERMQGPGQIQTHGIPPQMMGGPLQSSSSEGPQQNMWAARNDMPYPYQNR
- Purification:IgG purified
- Cat. No.:103401-676
Specifications
About this item
The ARID1B Antibody (2D2) from Novus Biologicals is a mouse monoclonal antibody to ARID1B. This antibody reacts with human. The ARID1B Antibody (2D2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ARID1B
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human