Anti-ARG2 Mouse Monoclonal Antibody [clone: 3D10]
Catalog # 103322-082
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ARG2
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:384
- Antigen Synonyms:L-arginine ureahydrolase|EC 3.5.3.1|Non-hepatic arginase|type II|arginase|mitochondrial|arginase-2|nonhepatic arginase|arginase 2|L-arginine amidinohydrolase|Type II arginase|Kidney-type arginase|EC 3.5.3|kidney arginase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ARG2 (AAH29050, 1 a.a. - 354 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSLRGSLSRLLQTRVHSILKKSVHSVAVIGAPFSQGQKRKGVEHGPAAIREAGLMKRLSSLGCHLKDFGDLSFTPVPKDDLYNNLIVNPRSVGLANQELAEVVSRAVSDGYSCVTLGGDHSLAIGTISGHARHCPDLCVVWVDAHADINTPLTTSSGNLHGQPVSFLLRELQDKVPQLPGFSWIKPCISSASIVYIGLRDVDPPEHFILKNYDIQYFSMRDIDRLGIQKVMERTFDLLIGKRQRPIHLSFDIDAFDPTLAPATGTPVVGGLTYREGMYIAEEIHNTGLLSALDLVEVNPQLATSEEEAKTTANLAVDVIASSFGQTREGGHIVYDQLPTPSSPDESENQARVRI
- Purification:IgG purified
- Cat. No.:103322-082
- Supplier no.:H00000384-M05
Specifications
About this item
The ARG2 Antibody (3D10) from Novus Biologicals is a mouse monoclonal antibody to ARG2. This antibody reacts with human. The ARG2 Antibody (3D10) has been validated for the following applications: ELISA, Immunoprecipitation.
Type: Primary
Antigen: ARG2
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human