Anti-APOA1 Mouse Monoclonal Antibody (DyLight 405) [clone: 1E12]
103405-236
- Antibody Type:Primary
- Antigen Name:Apolipoprotein A1
- Antigen Symbol:APOA1
- Clonality:Monoclonal
- Clone:1E12
- Conjugation:DyLight® 405
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:335
- Antigen Synonyms:ApoA-I|MGC117399|apolipoprotein A-I|Apo-AI|APOA1
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human Apolipoprotein A1 (25-267aa) purified from E. coli
- Purification:Protein G purified
- Cat. No.:103405-236
The Apolipoprotein A-I / ApoA1 Antibody (1E12) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to Apolipoprotein A-I / ApoA1. This antibody reacts with human. The Apolipoprotein A-I / ApoA1 Antibody (1E12) [DyLight 405] has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: 1E12
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human