Anti-APITD1 Mouse Polyclonal Antibody
Catalog # 103348-412
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:APITD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:378708
- Antigen Synonyms:TAF9-like domain 1|CENPSMHF1centromere protein S|FAAP16|Apoptosis-inducing TAF9-like domain-containing protein 1|apoptosis-inducing|CENP-SMGC32686|FANCM-interacting histone fold protein 1|Fanconi anemia-associated polypeptide of 16 kDa
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:APITD1 (NP_954988.1, 1 a.a. ~ 138 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103348-412
- Supplier no.:H00378708-B01P
Specifications
About this item
The APITD1 Antibody from Novus Biologicals is a mouse polyclonal antibody to APITD1. This antibody reacts with human. The APITD1 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: APITD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human