Anti-ANX4 Mouse Monoclonal Antibody [clone: 1D3]
Catalog # 103321-860
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Annexin A4
- Antigen Symbol:ANX4
- Clonality:Monoclonal
- Clone:1D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:307
- Antigen Synonyms:annexin-4|DKFZp686H02120|Chromobindin-4|Protein II|PIG28|proliferation-inducing gene 28|P32.5|ANX4Carbohydrate-binding protein p33/p41|proliferation-inducing protein 28|Annexin IV|annexin A4|Placental anticoagulant protein II|ZAP36|MGC75105|PP4-X|annexin IV (placental anticoagulant protein II)|Endonexin I|PAP-II|chromobindin-4,35-beta calcimedin|Lipocortin IV
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ANXA4 (AAH00182, 1 a.a. - 321 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAMATKGGTVKAASGFNAMEDAQTLRKAMKGLGTDEDAIISVLAYRNTAQRQEIRTAYKSTIGRDLIDDLKSELSGNFEQVIVGMMTPTVLYDVQELRRAMKGAGTDEGCLIEILASRTPEEIRRISQTYQQQYGRSLEDDIRSDTSFMFQRVLVSLSAGGRDEGNYLDDALVRQDAQDLYEAGEKKWGTDEVKFLTVLCSRNRNHLLHVFDEYKRISQKDIEQSIKSETSGSFEDALLAIVKCMRNKSAYFAEKLYKSMKGLGTDDNTLIRVMVSRAEIDMLDIRAHFKRLYGKSLYSFIKGDTSGDYRKVLLVLCGGDD
- Purification:IgG purified
- Cat. No.:103321-860
- Supplier no.:H00000307-M13
Specifications
About this item
The Annexin A4 Antibody (1D3) from Novus Biologicals is a mouse monoclonal antibody to Annexin A4. This antibody reacts with human. The Annexin A4 Antibody (1D3) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ANX4
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human