Anti-ANX2 Mouse Monoclonal Antibody [clone: 3E8-B6]
Catalog # 103321-842
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Annexin A2
- Antigen Symbol:ANX2
- Clonality:Monoclonal
- Clone:3E8-B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:302
- Antigen Synonyms:chromobindin 8|annexin-2|p36|LIP2PAP-IV|Lipocortin II|Protein I|Placental anticoagulant protein IV|ANX2L4LPC2|LPC2DP36|Calpactin I heavy chain|CAL1H|Calpactin-1 heavy chain|annexin A2|Annexin II|calpactin I heavy polypeptide|ANX2|Chromobindin-8
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ANXA2 (AAH09564.1, 1 a.a. - 339 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MFTVHEILCKLSLEGDHSTPPSAYGSVKAYTNFDAERDALNIETAIKTKGVDEVTIVNILTNRSNAQRQDIAFAYQRRTKKELASALKSALSGHLETLILGLLKTPAQYDASELKASMKGLGTDEDSLIEIICSRTNQELQEINRVYKEMYKTDLEKDIISDTSGDFRKLMVALAKGRRAEDGSVIDYELIDQDARDLYDAGVKRKGTDVPKWISIMTERSVPHLQKVFDRYKSYSPYDMLESIRKEVKGDLENAFLNLVQCIQNKPLYFADRLYDSMKGKGTRDKVLIRIMVSRSEVDMLKIRSEFKRKYGKSLYYYIQQDTKGDYQKALLYLCGGDD
- Purification:IgG purified
- Cat. No.:103321-842
- Supplier no.:H00000302-M01
Specifications
About this item
The Annexin A2 Antibody (3E8-B6) from Novus Biologicals is a mouse monoclonal antibody to Annexin A2. This antibody reacts with human. The Annexin A2 Antibody (3E8-B6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, RNA Inhibition.
Type: Primary
Antigen: ANX2
Clonality: Monoclonal
Clone: 3E8-B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human