Anti-ANP32B Rabbit Polyclonal Antibody
Catalog # 103358-944
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ANP32B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.025 mg
- Gene ID:10541
- Antigen Synonyms:PHAPI2SSP29|acidic (leucine-rich) nuclear phosphoprotein 32 family|Acidic protein rich in leucines|member B|acidic leucine-rich nuclear phosphoprotein 32 family member B|APRILSilver-stainable protein SSP29|Putative HLA-DR-associated protein I-2
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 1-17 of human APRIL.
- Purification:Protein G purified
- Cat. No.:103358-944
Specifications
About this item
The ANP32B Antibody from Novus Biologicals is a rabbit polyclonal antibody to ANP32B. This antibody reacts with human. The ANP32B Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ANP32B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human