Anti-AMPK gamma 1 Mouse Monoclonal Antibody [clone: 4A1G9]
103413-046
- Antibody Type:Primary
- Antigen Name:AMPK gamma 1
- Antigen Symbol:AMPK gamma 1
- Clonality:Monoclonal
- Clone:4A1G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:5571
- Antigen Synonyms:5'-AMP-activated protein kinase subunit gamma-1|gamma-1 subunit|MGC8666|5'-AMP-activated protein kinase|AMPK subunit gamma-1|protein kinase|AMPKg|AMP-activated|gamma 1 non-catalytic subunit|AMPK gamma1|AMPK gamma-1 chain
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Purified recombinant fragment of human PRKAG1 (AA: 230-331) expressed in E. Coli.
- Purification:Protein A or G purified
- Cat. No.:103413-046
The AMPK gamma 1 Antibody (4A1G9) from Novus Biologicals is a mouse monoclonal antibody to AMPK gamma 1. This antibody reacts with human. The AMPK gamma 1 Antibody (4A1G9) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
Type: Primary
Antigen: AMPK gamma 1
Clonality: Monoclonal
Clone: 4A1G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human