Anti-AMPK alpha 1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # 103413-042
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:AMPK alpha 1
- Antigen Symbol:AMPK alpha 1
- Clonality:Monoclonal
- Clone:2B7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Primates,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:5562
- Antigen Synonyms:alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1|catalytic alpha-1 chain|catalytic|alpha 1 catalytic subunit|AMP-activated protein kinase|AMPK|MGC33776|EC 2.7.11.1|protein kinase|MGC57364|AMPK subunit alpha-1|AMPK alpha 1,5'-AMP-activated protein kinase|AMP -activate kinase alpha 1 subunit|EC 2.7.11|AMPK1|AMPKa1|AMP-activated
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Purified recombinant fragment of human AMPK alpha 1 expressed in E. coli. [UniProt# Q13131]
- Purification:Ammonium sulfate precipitation
- Cat. No.:103413-042
Specifications
About this item
The AMPK alpha 1 Antibody (2B7) from Novus Biologicals is a mouse monoclonal antibody to AMPK alpha 1. This antibody reacts with human, mouse, rat, primate. The AMPK alpha 1 Antibody (2B7) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Simple Western ,
Type: Primary
Antigen: AMPK alpha 1
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primate