Anti-ALDC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4A9]
Catalog # 103409-788
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Aldolase C
- Antigen Symbol:ALDC
- Clonality:Monoclonal
- Clone:4A9
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine
- Western Blot:Yes
- Size:100 μl
- Gene ID:230
- Antigen Synonyms:fructose-1,6-biphosphate triosephosphate lyase|aldolase 3|fructose-bisphosphate|Brain-type aldolase|ALDC|fructose-bisphosphate aldolase C|fructoaldolase C|aldolase C|EC 4.1.2.13
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The N-terminal 20 amino acids of human Aldolase C (MPHSYPALSAEQKKELSDIA). [UniProt# P09972]
- Purification:Protein G purified
- Cat. No.:103409-788
Specifications
About this item
The Aldolase C Antibody (4A9) [HRP] from Novus Biologicals is a mouse monoclonal antibody to Aldolase C. This antibody reacts with human, bovine. The Aldolase C Antibody (4A9) [HRP] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ALDC
Clonality: Monoclonal
Clone: 4A9
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine