Anti-ALDC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4A9]
Catalog # 103409-786
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Aldolase C
- Antigen Symbol:ALDC
- Clonality:Monoclonal
- Clone:4A9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine
- Size:100 μl
- Gene ID:230
- Antigen Synonyms:fructose-1,6-biphosphate triosephosphate lyase|aldolase 3|fructose-bisphosphate|Brain-type aldolase|ALDC|fructose-bisphosphate aldolase C|fructoaldolase C|aldolase C|EC 4.1.2.13
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The N-terminal 20 amino acids of human Aldolase C (MPHSYPALSAEQKKELSDIA). [UniProt# P09972]
- Purification:Protein G purified
- Cat. No.:103409-786
Specifications
About this item
The Aldolase C Antibody (4A9) [FITC] from Novus Biologicals is a mouse monoclonal antibody to Aldolase C. This antibody reacts with human, bovine. The Aldolase C Antibody (4A9) [FITC] has been validated for the following applications: Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: ALDC
Clonality: Monoclonal
Clone: 4A9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine