Anti-AGPAT9 Rabbit Polyclonal Antibody
Catalog # 103284-958
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:AGPAT9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:84803
- Antigen Synonyms:GPAT3|GPAT-3,1-acylglycerol-3-phosphate O-acyltransferase 8|hGPAT3|1-acylglycerol-3-phosphate O-acyltransferase 9|theta|lysophosphatidic acid acyltransferase theta|MGC11324|MAG-1|HMFN0839,1-AGP acyltransferase 9|MAG1|UNQ2753/PRO6492|glycerol-3-phosphate acyltransferase 3|acyl-CoA:glycerol-3-phosphate acyltransferase 3
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:ILVKTLEWATIRIEKGTPKESILKNSASVGIIQRDESPMEKGLSGLRGRDFELSDVFYFSKKGLEAIVEDEVTQRFSSEELVSWNLLTRTN
- Purification:Immunogen affinity purified
- Cat. No.:103284-958
- Supplier no.:NBP1-93692
Specifications
About this item
The AGPAT9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AGPAT9. This antibody reacts with human. The AGPAT9 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: AGPAT9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human