Anti-ADAMTSL5 Rabbit Polyclonal Antibody
Catalog # 103284-700
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ADAMTSL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:339366
- Antigen Synonyms:domain containing 6|thrombospondin type-1 domain-containing protein 6|ADAMTS-like 5|thrombospondin|ADAMTS-like protein 5|ADAMTSL-5|THSD6|type I
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:AHRLLHYCGSDFVFQARVLGHHHQAQETRYEVRIQLVYKNRSPLRAREYVWAPGHCPCPMLAPHRDYLMAVQRLVSPDGTQDQ
- Purification:Immunogen affinity purified
- Cat. No.:103284-700
- Supplier no.:NBP1-93438
Specifications
About this item
The ADAMTSL5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ADAMTSL5. This antibody reacts with human. The ADAMTSL5 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ADAMTSL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human