To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ADAMTS2
- Clonality:Monoclonal
- Clone:7G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μg
- Gene ID:9509
- Antigen Synonyms:ADAM-TS 2|ADAMTS-2|pNPI|A disintegrin and metalloproteinase with thrombospondin motifs 2|Procollagen N-endopeptidase|Procollagen I N-proteinase|PCINPhPCPNI|ADAM-TS2EC 3.4.24.14|PCPNI|a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif|ADAM metallopeptidase with thrombospondin type 1 motif|Procollagen I/II amino propeptide-processing enzyme|EC 3.4.24|2|NPIDKFZp686F12218|ADAMTS-3|PC I-NP|PCI-NP
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ADAMTS2 (NP_055059, 1112 a.a. ~ 1211 a.a) partial recombinant protein with GST tag.KHNDIDVFMPTLPVPTVAMEVRPSPSTPLEVPLNASSTNATEDHPETNAVDEPYKIHGLEDEVQPPNLIPRRPSPYEKTRNQRIQELIDEMRKKEMLGK
- Purification:IgG purified
- Cat. No.:103337-124
- Supplier no.:H00009509-M03
Specifications
About this item
The ADAMTS2 Antibody (7G3) from Novus Biologicals is a mouse monoclonal antibody to ADAMTS2. This antibody reacts with human, rat. The ADAMTS2 Antibody (7G3) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: ADAMTS2
Clonality: Monoclonal
Clone: 7G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat