Anti-ACTRIIB Mouse Polyclonal Antibody
Catalog # 103321-376
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Activin RIIB
- Antigen Symbol:ACTRIIB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 ug
- Gene ID:93
- Antigen Synonyms:activin A receptor|ActR-IIB|Activin receptor type IIB|MGC116908|type IIB|ACTRIIB|EC 2.7.11.30|EC 2.7.11|activin receptor type-2B
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ACVR2B (AAH96245.1, 1 a.a. - 303 a.a.) full-length human protein. MNDFVAVKIFPLQDKQSWQSEREIFSTPGMKHENLLQFIAAEKRGSNLEVELWLITAFHDKGSLTDYLKGNIITWNELCHVAETMSRGLSYLHEDVPWCRGEGHKPSIAHRDFKSKNVLLKSDLTAVLADFGLAVRFEPGKPPGDTHGQVGTRRYMAPEVLEGAINFQRDAFLRIDMYAMGLVLWELVSRCKAADGPVDEYMLPFEEEIGQHPSLEELQEVVVHKKMRPTIKDHWLKHPGLAQLCVTIEECWDHDAEARLSAGCVEERVSLIRRSVNGTTSDCLVSLVTSVTNVDLPPKESSI
- Purification:Protein G purified
- Cat. No.:103321-376
- Supplier no.:H00000093-B01P
Specifications
About this item
The Activin RIIB Antibody from Novus Biologicals is a mouse polyclonal antibody to Activin RIIB. This antibody reacts with human. The Activin RIIB Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ACTRIIB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human