Anti-5-HT7 Rabbit Polyclonal Antibody
Catalog # 103358-896
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:5-HT7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:0.025 mg
- Gene ID:3363
- Antigen Synonyms:5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled)|5-HT75-hydroxytryptamine receptor 7|Serotonin receptor 7|5-HT-7|serotonin 5-HT-7 receptor|5-HT-X
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 13-28 of the rat 5-HT7R (AAA42134.1).
- Purification:Protein G purified
- Cat. No.:103358-896
Specifications
About this item
The 5-HT7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to 5-HT7. This antibody reacts with human, mouse, rat, canine. The 5-HT7 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: 5-HT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine