ContactUsLinkComponent
Unger® Opti-Loc Extension Pole
Unger® Opti-Loc Extension Pole
  102866-074
undefined
Unger® Opti-Loc Extension Pole
  102866-074
 :  UNGED900
  • Description:
    Aluminum Extension Pole, Three Sections, Green/Silver
  • Length:
    30'
  • Cat. No.:
    102866-074
  • Supplier no.:
    UNGED900
Anodized, extruded aluminum pole with nylon locking collar.
  • Snap-in threaded locking cone makes it easy to add and remove tools, even when wet
  • Accepts tools with tapered or threaded sockets
  • Ergonomically designed nylon handle grip for comfort
  • Poles are sectioned for convenient carrying and storage
Aluminum handle. Green/silver color.