Metric O-Ring Assortment, Precision Brand
Catalog # 300049-539
Supplier: Safety & Industrial Supplies
Specifications
- Description:Metric O-Ring Assortment
- Material:Buna-N
- Cat. No.:300049-539
Specifications
About this item
Made of BUNA-A 70 durometer rubber compound, these metric size rings create a tight seal for fluid and gas connections.
Delivery: Set includes (10) 2.5 mm Body, 17 mm, (10) 2.5 mm Body, 19 mm, (10) 2 mm Body, 14 mm, (10) 2 mm Body, 20 mm, (10) 3 mm Body, 25 mm, (10) 3 mm Body, 27 mm, (10) 3 mm Body, 28 mm, (10) 3 mm Body, 30 mm, (15) 2.5 mm Body 16 mm, (15) 2.5 mm Body, 10 mm, (15) 2.5 mm Body, 12 mm, (15) 2.5 mm Body, 14 mm, (15) 2.5 mm Body, 15 mm, (15) 2 mm Body 10 mm, (15) 2 mm Body 6 mm, (15) 2 mm Body, 12 mm, (15) 2 mm Body, 3 mm, (15) 2 mm Body, 4 mm, (15) 2 mm Body, 5 mm, (15) 2 mm Body, 8 mm, (15) 3 mm Body, 19 mm, (15) 3 mm Body, 20 mm, (15) 3 mm Body, 22 mm, (15) 3 mm Body, 24 mm.