Order Entry
United States
ContactUsLinkComponent
 
Crystal Simple Green, Simple Green®
  300009-878
undefined
Crystal Simple Green, Simple Green®
  300009-878
 :  676-19005

 

  • Cat. No.:
    300009-878
  • Packaging:
    Pail
  • pH:
    9.35
  • Specific gravity:
    1.02
  • Volume:
    5 gal.
  • Supplier no.:
    676-19005

 

 

Effective concentrated cleaner and degreaser for industrial and manufacturing applications

  • Suitable for critical cleaning tasks that demand a fragrance-free solution
  • Clear, unscented formulation

Use to remove grease, grime, lubricants, and oil-based soil on ceramics, cement/concrete, enamel, fiberglass, laminates, metal, and plastic.

High rinsability factor meets requirements for cleaning in cosmetic, medical, and pharmaceutical environments.