Proto® Blade Long Feeler Gauge Set, 25, Stanley® Products
Catalog # 300009-472
Supplier: Safety & Industrial Supplies
Specifications
- Description:25 Blade Long Feeler Gauge Set, incl. Steel Holder
- Dimensions:12 Lx0.5" W
- Environmentally Preferable:
- Cat. No.:300009-472
- Supplier no.:577-000TL
Specifications
About this item
Convenient blades are etched with both decimal size and metric equivalents for easy measurement.
- Easily measures gap widths
- Durable, flexible steel construction
Delivery: Set includes 25 blades of different thickness such as 0.0015, 0.002, 0.003, 0.004, 0.005, 0.006, 0.008, 0.009, 0.01, 0.011, 0.012, 0.013, 0.014, 0.015, 0.016, 0.017, 0.018, 0.019, 0.02, 0.021, 0.025, 0.027, 0.03, and 0.04.