Extended Performance Type 27 Depressed Center Wheel, DeWalt
Catalog # 300077-087
Supplier: Safety & Industrial Supplies
Specifications
- Material:Silicon carbide
- Cat. No.:300077-087
Specifications
About this item
Use to cut/notch ferrous metal and stainless steel.
- Z24R grit
- 0.625 – 11" arbor thread
- 4.5" diameter and 0.125" thickness
Speed range of 13300 rpm.