Combined Eosinophil - Mast Cell (C.E.M.) Stain Kit, Azer Scientific
10837-234
:
- Description:Combined Eosinophil - Mast Cell Stain Kit
- Cat. No.:10837-234
- Supplier no.:ES4805
The C.E.M. stain kit demonstrates eosinophils and mast cells in fixed or frozen tissue sections.
- For in-vitro diagnostic use only
- Pre-mixed and ready to use
Combined eosinophil - mast cell stain kit will stain mast cells and eosinophils in tissue sections or any air dried smear applied to a microscope slide. Mast cells appear bright blue, eosinophils stain bright red and nucleai stain blue.
: The complete kit includes 125 ml of astra blue solution, 125 ml of vital new red solution and 125 ml of Mayer's hematoxylin (Lillie's modified).
: Use in chemical fume hood whenever possible.