Order Entry
United States
ContactUsLinkComponent
Specimen Container, Azer Scientific
Specimen Container, Azer Scientific
Catalog # 95057-962
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Specimen Container, Azer Scientific
Catalog # 95057-962
Supplier Number:  ES3408
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Sterility:
    Non sterile
  • Capacity:
    20 ml
  • Description:
    Specimen Container
  • Cat. No.:
    95057-962
  • Supplier no.:
    ES3408

Specifications

About this item

This 20 mL specimen container is supplied without a lid.

Packaging: 1000/case.