ContactUsLinkComponent
Dykem™ Brite-Mark™ Medium Markers, ORS Nasco
Dykem™ Brite-Mark™ Medium Markers, ORS Nasco
  300000-769
undefined
Dykem™ Brite-Mark™ Medium Markers, ORS Nasco
  300000-769
 :  253-84002
  • Tip type:
    Medium bullet
  • Color:
    Black
  • Cat. No.:
    300000-769
  • Supplier no.:
    253-84002
Bullet tips are replaceable to assure complete use of paint in each marker
  • Drying time is 25 – 35 seconds
  • Heavy-duty barrel retains its shape
  • Ideal for color coding
Makes permanent bright marks on metal, rubber, plastic, glass, concrete, leather, stone, wood, vinyl, and more.
Valve action mechanism prevents paint in barrel from drying when cap is left off.