- Antibody Type:Primary
- Antigen Name:keratin 4, keratin 5, keratin 6A, keratin 6B, keratin 8, keratin 10, keratin 13, keratin 18
- Antigen Symbol:KRT4, KRT5, KRT6A, KRT6B, KRT8, KRT10, KRT13, KRT18
- Clonality:Monoclonal
- Clone:SPM583
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Marmoset,Frog,Pig,Goat,Human,Bovine,Guinea pig,Rat,Mouse
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P19013, P13647, P02538, P04259, P48668, P05787, P13645, P13646, P05783
- Antigen Synonyms:KRT4, KRT5, KRT6A, KRT6B, KRT8, KRT10, KRT13, KRT18
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:A keratin-enriched preparation from cultured human A431 cells was used as the immunogen for this anti-Multi-Cytokeratin antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-388
This mAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1 ug/ml Western blot: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the anti-Multi-Cytokeratin antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: KRT4, KRT5, KRT6A, KRT6B, KRT8, KRT10, KRT13, KRT18
Clonality: Monoclonal
Clone: SPM583
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Rat, Mouse, Guinea Pig, Frog, Goat, Marmoset, Pig