Specifications
- Antibody Type:Primary
- Antigen Name:mucin 3A, cell surface associated
- Antigen Symbol:MUC3A
- Clonality:Monoclonal
- Clone:MUCN3-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q02505
- Antigen Synonyms:MUC3A
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Amino acids (C-HSTPSFTSSITTTETTSHSTPSFTSSITTTETTS) which contains two of the MUC3 tandem repeats was used as the immunogen for the MUC3 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-992
Specifications
About this item
The Mucins are a family of highly glycosylated, secreted proteins with a basic structure consisting of a variable number of tandem repeats. Mucin-3/MUC3 is a major glycoprotein component of a variety of mucus gels. Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces. May be involved in ligand binding and intracellular signaling. [UniProt]
Immunofluorescence: 1-2 ug/ml Immunohistochemistry (FFPE): 1-2 ug/ml for 30 min at RT (1) Prediluted format : incubate for 30 min at RT (2) Optimal dilution of the MUC3 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 min
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: MUC3A
Clonality: Monoclonal
Clone: MUCN3-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human