Order Entry
United States
ContactUsLinkComponent
 
Anti-MITF Mouse Monoclonal Antibody [clone: MPAF3]
Catalog # 76194-976
Supplier:  Prosci
Anti-MITF Mouse Monoclonal Antibody [clone: MPAF3]
Catalog # 76194-976
Supplier:  Prosci
Supplier Number:  33-949

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    microphthalmia-associated transcription factor
  • Antigen Symbol:
    MITF
  • Clonality:
    Monoclonal
  • Clone:
    MPAF3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    100 ug
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    O75030
  • Antigen Synonyms:
    MITF
  • Storage Buffer:
    PBS with 0.1 mg/ml BSA and 0.05% sodium azide
  • Storage Temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Recombinant full-length human protein was used as the immunogen for the MITF antibody.
  • Purification:
    Protein G affinity chromatography
  • Cat. No.:
    76194-976

Specifications

About this item

MITF (microphthalmia transcription factor) is a transcription factor that regulates the expression of genes with essential roles in cell differentiation, proliferation and survival. Binds to symmetrical DNA sequences (E-boxes) (5'-CACGTG-3') found in the promoters of target genes, such as BCL2 and tyrosinase (TYR). Plays an important role in melanocyte development by regulating the expression of tyrosinase (TYR) and tyrosinase-related protein 1 (TYRP1). Plays a critical role in the differentiation of various cell types, such as neural crest-derived melanocytes, mast cells, osteoclasts and optic cup-derived retinal pigment epithelium. [UniProt]

Immunofluorescence: 1-2 ug/ml Immunohistochemistry (FFPE): 1-2 ug/ml for 30 min at RT (1) Prediluted format : incubate for 30 min at RT (2) Titering of the MITF antibody may be required for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: MITF
Clonality: Monoclonal
Clone: MPAF3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human