Specifications
- Antibody Type:Primary
- Antigen Name:F-box only protein 5
- Antigen Symbol:FBXO5
- Clonality:Monoclonal
- Clone:FBXP5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q9UKT4
- Antigen Synonyms:FBXO5
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:A human partial recombinant N-terminal protein fragment was used as the immunogen for the EMI-1 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-942
Specifications
About this item
Early Mitotic Inhibitor-1 regulates mitosis by inhibiting the anaphase promoting complex/cyclosome (APC). It is a conserved F box protein containing a zinc-binding region essential for APC inhibition. The protein functions to promote cyclin A accumulation and S phase entry in somatic cells by inhibiting the APC complex. At the G1-S transition, EMI-1 is transcriptionally induced by the E2F transcription factor. Overexpression accelerates S-phase entry and can override a G1 block caused by overexpression of Cdh1 or the E2F-inhibitor p105 retinoblastoma protein (pRb). Depleting cells of EMI-1 through RNA interference prevents accumulation of cyclin A and inhibits S phase entry.
Western blot: 1-2 ug/ml Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) Optimal dilution of the EMI-1 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0 for 10-20 min followed by cooling at RT for 20 minutes
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: FBXO5
Clonality: Monoclonal
Clone: FBXP5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human