Specifications
- Antibody Type:Primary
- Antigen Name:tumor protein p53
- Antigen Symbol:TP53
- Clonality:Monoclonal
- Clone:TP53/1799R
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P04637
- Antigen Synonyms:TP53
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human p53 protein was used as the immunogen for this recombinant p53 antibody.
- Purification:Protein A affinity chromatography
- Cat. No.:76194-824
Specifications
About this item
Recognizes a 53 kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53. p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to MDM2, SV40 T antigen and human papilloma virus E6 protein. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia. Mutations involving p53 are found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1 ug/ml Western blot: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.25-0.5 ug/ml for 30 min at RT Prediluted IHC only format: incubate for 30 min at RT (1) The optimal dilution of the recombinant p53 antibody for each application should be determined by the researcher.
1. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: TP53/1799R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG, kappa
Reactivity: Human