- Antibody Type:Primary
- Antigen Name:myosin heavy chain 11
- Antigen Symbol:MYH11
- Clonality:Monoclonal
- Clone:SMMS-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Rabbit,Chicken,Human,Bovine,Guinea pig,Dog,Rat,Cat
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P35749
- Antigen Synonyms:MYH11
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human uterus extract was used as the immunogen for the SM-MHC antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-174
Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The stated application concentrations are suggested starting amounts. Optimal dilution of the SM-MHC antibody should be determined by the researcher.
1. Staining of formalin/paraffin tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 min.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: MYH11
Clonality: Monoclonal
Clone: SMMS-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Bovine, Pig, Dog, Cat, Rabbit, Rat, Guinea Pig, Chicken